missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ WFDC1 Polyclonal Antibody
GREENER_CHOICE

Produktkod. 17710914
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17710914 200 μL
17752894 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17710914 Leverantör Invitrogen™ Leverantörsnummer PA5119507

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

WFDC1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

This gene encodes a member of the WAP-type four disulfide core domain family. The WAP-type four-disulfide core domain, or WAP signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor in many family members. The encoded protein shares 81% amino acid identity with the rat ps20 protein, which was originally identified as a secreted growth inhibitor. This gene is mapped to chromosome 16q24, an area of frequent loss of heterozygosity in cancers, including prostate, breast and hepatocellular cancers and Wilms' tumor. Owing to its location and a possible growth inhibitory property of its gene product, this gene is suggested to be a tumor suppressor gene.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen WFDC1
Användningsområden Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Klassificering Polyclonal
Koncentration 0.5 mg/mL
Konjugera Unconjugated
Formulering PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gen WFDC1
Genaccessionsnr. Q9ESH5, Q9HC57
Gene Alias 2310058A03Rik; Prostate stromal protein ps20; PS20; ps20 growth inhibitor; WAP four-disulfide core domain 1; WAP four-disulfide core domain 1 homolog; WAP four-disulfide core domain protein 1; Wfdc1
Gensymboler WFDC1
Värd art Rabbit
Immunogen Recombinant protein WAP Four Disulfide Core Domain Protein 1. The antigen corresponds to amino acid range 24-208 of the target protein.
Reningsmetod Antigen affinity chromatography, Protein A
Kvantitet 200 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 58189, 67866
Målarter Human, Mouse
Innehåll och lagring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.