Learn More
WFS1 Mouse anti-Human, Unconjugated, Clone: 4C2B5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67291-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The WFS1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human WFS1. This antibody recognizes human, mouse, rat, pig antigen. The WFS1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
WFS1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, ELISA | |
4C2B5 | |
Unconjugated | |
150 μL | |
7466 | |
IgG1 |