Learn More
Cytiva Whatman™ Grade 54 Cellulose Chromatography Paper
Product Code. 11303025
Change view
Click to view available options
Length (Metric):
91.4 m
Unit Size:
Each
Product Code. Length (Metric) Unit Size
11303025 91.4 m Each
1 options
Product Code. 11303025 Supplier Cytiva Supplier No. 3454651
  • ONLINE EXCLUSIVE PRICE
    2080.00 SEK
    1891.00 SEK / Paket med 1
    Save 189.00 SEK
    9% Av
Beräknad leverans: 14-04-2027
54 SFC cellulose chromatography roll is a 0.18 hardened paper for routine chromatography.
  • Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind.
  • Manufactured and tested specifically for chromatographic techniques. This ensures the wicking capability and uniformity of capillary action that are important in chemical separations.
  • Also widely used in protein and nucleic acid blotting.
TRUSTED_SUSTAINABILITY
Specifications
Description 54 SFC cellulose chromatography roll is a 0.18 hardened paper for routine chromatography.
Dimensions (L x W) 38.1 cm x 91.44 m (1.5 in. x 300 ft.)
Film Thickness 0.18 mm
For Use With (Application) For Protein and Nucleic Acid Blotting
Format Roll
Grade Grade 54 SFC
Material Cellulose
Width (English) 1.5 in.
Width (Metric) 3.8 cm
Flow Rate 180 mm/30 min.
Length (Metric) 91.4 m