Learn More
Cytiva Whatman™ Grade 8 Ruled Special-Purpose Filter Paper
Hardened grade for routine analysis of extraneous matter in food, beverage and pharmaceutical laboratories
Brand:  Cytiva 10347009
Additional Details : Weight : 0.06000kg
 View more versions of this product
Product Code. 11969337
  • 515.00 SEK / Pack of 100
Estimated Shipment: 14-05-2024
to see stock.
Add to basket
Description
Description
™Resistant to aqueous and non-aqueous solutions
Recovers and measures contaminants in food products™
Specifications
Specifications
Cotton Linters
5mm green rules are resistant to aqueous, non-aqueous solution
3.54 in.
Qualitative analytical techniques to determine and identify materials
Ruled Special Purpose Filter Paper
9 cm