Learn More
Cytiva™ Whatman™ Nuclepore Hydrophilic Membrane
Product Code. 16655665
Change view
Click to view available options
Pore Size:
1 μm
2 μm
3 μm
8 μm
10 μm
12 μm
Diameter (Metric):
13 mm
19 mm
25 mm
47 mm
90 mm
293 mm
Unit Size:
Pack of 100
Pack of 25
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Diameter (Metric) Unit Size
16655665 3 μm 25 mm Pack of 100
16675675 1 μm 13 mm Pack of 100
16685675 1 μm 19 mm Pack of 100
16605685 1 μm 47 mm Pack of 100
16311426 8 μm 25 mm Pack of 100
16321426 8 μm 47 mm Pack of 100
16675665 10 μm 13 mm Pack of 100
16685665 10 μm 25 mm Pack of 100
16695665 10 μm 47 mm Pack of 100
16615675 12 μm 13 mm Pack of 100
16625675 12 μm 25 mm Pack of 100
16635675 12 μm 47 mm Pack of 100
16615685 1 μm 90 mm Pack of 25
16635685 1 μm 293 mm Pack of 25
16645665 3 μm 13 mm Pack of 100
16645685 2 μm 25 mm Pack of 100
16655685 2 μm 47 mm Pack of 100
16665665 3 μm 47 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Product Code. 16655665 Supplier Cytiva™ Supplier No. 10418306
  • ONLINE EXCLUSIVE PRICE
    937.00 SEK
    847.87 SEK / Paket med 100
    Save 89.13 SEK
    9% Av
I lager
Nuclepore track-etched hydrophilic membranes with highly defined pore size cutoff in sub-micron range
  • Track-etched for highly defined pore size and high pore density
  • Treated with PVP to make it hydrophilic
  • Low extractables minimizing filtration sample contamination risk
  • High chemical resistance
  • Good thermal stability
  • Low, consistent ash
  • Low tare weights
  • Smooth, flat membrane surface for high particle visibility
Applications:
  • Bioassays
  • Cell biology
  • Environmental analysis
  • Microscopy
  • Cytology
  • Parasitology
  • Certain air and oceanography analyses
TRUSTED_SUSTAINABILITY
Specifications
Material Polycarbonate
Pore Size 3 μm
For Use With (Application) Bioassays, Cell biology, Environmental analysis, Microscopy, Cytology, Parasitology, Certain air and oceanography analyses
Quantity 100 Pk.
Type Nuclepore hydrophilic membrane
Diameter (Metric) 25 mm
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.