missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Sugar/Food Industry Filter Papers

Used for clarifying filtration of beet juice/pulp. Whatman™ Grade 300 Sugar Industry Filter Papers are smooth cellulose filter papers with good particle retention capabilities at a relatively high filtration speed.

Brand:  Cytiva 10316114

Additional Details : Weight : 1.71900kg

Product Code. 11378894

  • 1530.00 SEK / Pack of 1000
Estimated Shipment: 24-06-2024
to see stock.
Add to basket

Description

Description

Used for the clarifying filtration of:

  • Dried beet pulp extracts
  • Beet juice after the addition of lead acetate for subsequent polarimetric sugar determination
Specifications

Specifications

68 g/m2
Disc
95 s (Herzberg)
3000
160 μm
Smooth
Cellulose
Clarifying filtration of beet juice/pulp extracts in the sugar industry
1000Pack
185 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers