Learn More
DWK Life Sciences Wheaton™ Aluminum Seals: Flip Cap
Flip Cap Seals are easy to use, autoclavable and tamper-evident
Brand:  DWK Life Sciences 224202
Additional Details : Weight : 0.54430kg
Product Code. 10708404
  • 2345.00 SEK / Pack of 1000
Estimated Shipment: 31-05-2024
to see stock.
Add to basket
You may only order up to the available stock. For more details call customer services on +46 31 352 32 00.
Description
Description
Flip Cap Seals are easy to use, autoclavable and tamper-evident
  • The red plastic top flips off easily with pressure from the thumb exposing the aluminum seal with center hole and pull ring
  • For Vials with a 13mm OD