Learn More
DWK Life Sciences Wheaton™ Black Phenolic Cap with Rubber Liner
Click to view available options
Neck Finish:
28-400
33-400
43-400
48-400
Specifications
Closure Liner | Rubber |
For Use With (Equipment) | Bottle |
Color | Black |
Material | Phenolic Resin |
No. per Case | 1000 Cs. |
Neck Finish | 33-400 |
Certifications/Compliance | ISO 9001 |