Learn More
DWK Life Sciences Wheaton™ Clear Boston Rounds with PE Cone-Lined Caps
Catalog Number 12389729
Click to view available options
Capacity:
1 oz., 30mL
16 oz., 500 mL
2 oz., 60 mL
32 oz., 1000 mL
4 oz., 125 mL
8 oz., 250 mL
Diameter (English):
1.22 in.
1.53 in.
1.88 in.
2.36 in.
2.95 in.
3.7 in.
Height (English):
3.1 in.
3.7 in.
4.4 in.
5.43 in.
6.6 in.
8.18 in.
Ideal for solvent, chemical and sample storage
  • Well suited for storage and sampling of liquids; clear Type III soda-lime glass provides visibility of contents
  • Narrow mouth enables liquids to be carefully poured to prevent spillage
Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage.
Order Info
Caps can be ordered separately.
Capacity 2 oz., 60 mL
Closure Liner Polyethylene
Closure Material Phenolic
Closure Size 20-400 mm
Closure Type Screw Cap
Diameter (Metric) 39 mm
Height (English) 3.7 in.
Height (Metric) 94 mm
Capacity (Metric) 60 mL
Capacity (English) 2 oz.
Color Clear
Diameter (English) 1.53 in.
Material Soda Lime Glass
Closure Color Black
Certifications/Compliance EPA, FDA