Learn More
DWK Life Sciences Wheaton™ Clear French Squares with Polyvinyl-lined Closures
Catalog Number 12309809
Click to view available options
Capacity:
0.5 oz., 15 mL
1 oz., 30 mL
1 oz., 30mL
16 oz., 500 mL
16.9 oz., 500 mL
2 oz., 60 mL
2.03 oz., 60 mL
4 oz., 125 mL
4.23 oz., 125 mL
8 oz., 250 mL
Diameter (English):
1.2 in.
1.22 in.
1.53 in.
1.77 in.
2.2 in.
2.67 in.
Height (English):
2.52 in.
2.91 in.
3.5 in.
4.44 in.
5.47 in.
6.65 in.
Square shape maximizes lab storage space
  • USP Type III soda-lime glass
  • Ideal for solvent, chemical or sample storage
Capacity 1 oz., 30 mL
Closure Liner Polyvinyl
Closure Material Polypropylene
Closure Size 24-400 mm
Closure Type Screw Cap
Diameter (Metric) 31 mm
Height (English) 2.91 in.
Height (Metric) 74 mm
Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Clear
Diameter (English) 1.22 in.
Material Soda Lime Glass
Closure Color White