Läs mer
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Vials
Handla allt Wheaton Science Products Produkter
Färg:
Klart
Volym (metrisk):
2 ml
5 ml
10 ml
Förpackningsstorlek:
Paket med 158
Beskrivning
- Sterilized, off-the-shelf, ready-to-use (RtU) products
- Variety of Kitted and stand-alone sterile components – a solution for any application
- Complete manufacturing & certificate of analysis traceability
- USP 788/EP 2.9.19 Particulate Matter in Injections, USP 85/EP 2.6.1, The Bacterial Endotoxins Test and USP 71/ EP 2.6.14 The Sterility Test, USP 660/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP 381/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
- Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
- Amber versions for light sensitive content packaging
Certifikat
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Specifikationer
Specifikationer
| Färg | Klart |
| Material | Borosilicate Glass (Type 1) |
| Sterilitet | Sterile |
| Volym (metrisk) | 2 ml |
| nr per ärende | 210 |
| Produktlinje | Wheaton CompletePAK |
| Certifikat och efterlevnad | USP 788, 85, and 71, Container Closure Integrity (CCI) certified |