Learn More
DWK Life Sciences WHEATON™ Culture Vessel
Product Code. 12438952 Shop All Wheaton Science Products Products
Change view
Click to view available options
Height (Metric):
355 mm
535 mm
Unit Size:
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
12438952 355 mm Pack of 4
12448952 535 mm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12438952 Supplier DWK Life Sciences Supplier No. 348524
  • ONLINE EXCLUSIVE PRICE
    6995.00 SEK
    6025.55 SEK / Paket med 4
    Save 969.45 SEK
    13% Av
Beräknad leverans: 14-04-2027
WHEATON™ Culture Vessel is designed for repeated wet or dry sterilization and is made of borosilicate glass that conforms to USP and ASTM requirements. Features a 2.6mm wall thickness and screw head closure for reliable, reusable cell culture applications.
  • Conforms to USP and ASTM requirements
  • Made of Type I borosilicate glass
  • 2.6mm wall thickness for durability
  • 38, 45, or 51mm screw head closure
  • Autoclavable — suitable for repeated wet or dry sterilization
  • Reusable design for cost-effective cell culture workflows
Specifications
Capacity (Metric) 2500 mL
Color Clear
Material Borosilicate Glass
Closure Included Yes
Mouth Narrow
Height (Metric) 355 mm
Diameter (Metric) 110 mm
Autoclavable Autoclavable
Shape Cylindrical
Product Type Roller Bottle