Learn More
DWK Life Sciences WHEATON™ Culture Vessel
Product Code. 12448952 Shop All DWK Life Sciences Products
Change view
Click to view available options
Height (Metric):
355 mm
535 mm
Unit Size:
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
12448952 535 mm Pack of 4
12438952 355 mm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12448952 Supplier DWK Life Sciences Supplier No. 348528
  • ONLINE EXCLUSIVE PRICE
    10440.00 SEK
    7719.22 SEK / Paket med 4
    Save 2720.78 SEK
    26% Av
Beräknad leverans: 21-04-2027
WHEATON™ Culture Vessel is designed for repeated wet or dry sterilization and is made of borosilicate glass that conforms to USP and ASTM requirements. Features a 2.6mm wall thickness and screw head closure for reliable, reusable cell culture applications.
  • Conforms to USP and ASTM requirements
  • Made of Type I borosilicate glass
  • 2.6mm wall thickness for durability
  • 38, 45, or 51mm screw head closure
  • Autoclavable — suitable for repeated wet or dry sterilization
  • Reusable design for cost-effective cell culture workflows
Specifications
Capacity (Metric) 4229 mL
Color Clear
Material Borosilicate Glass
Closure Included Yes
Height (Metric) 535 mm
Diameter (Metric) 110 mm
Autoclavable Autoclavable
Capacity (English) 143 oz.
Shape Round
Product Type Roller Bottle