Learn More
DWK Life Sciences Wheaton™ Grinding Chamber Replacement Parts For Tapered Tissue Grinders With PTFE Pestles
Tapered tissue grinders replacement grinding chamber for fluoropolymer resin pestle
1585.00 SEK - 1860.00 SEK
Specifications
Material | Borosilicate Glass |
---|---|
Product Type | Grinding Chamber |
For Use With (Application) | Tapered tissue grinder with PTFE pestle |
For Use With (Equipment) | Tapered tissue grinder |
Product Code | Brand | Capacity | Length (English) Tube | Tube Diameter (English) Outside | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Length (English) Tube | Tube Diameter (English) Outside | Price | Quantity & Availability | |||
11758272
|
DWK Life Sciences
358132 |
0.03 oz., 1mL | 1.9 in. | 0.43 in. |
1585.00 SEK
Pack of 2 |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||
11738282
|
DWK Life Sciences
358140 |
0.16 oz., 5mL | 3.6 in. | 0.51 in. |
1860.00 SEK
Pack of 2 |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||
11758282
|
DWK Life Sciences
358144 |
0.5 oz., 15mL | 4.48 in. | 0.7 in. |
1740.00 SEK
Pack of 2 |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||
Description
- Grinders are available with glass or fluoropolymer resin pestles
- Glass pestles are ideal for the homogenization of heart, muscle, lung, skin, and plant tissue
- Fluoropolymer resin pestles are ideal for use with soft tissues such as brain and feature a stainless steel shaft with a 6.3mm (1/4 in.) diameter
- Grinding chamber is made of low extractable borosilicate glass
- The conical tip of the grinding chamber allows initial reduction, followed by passage through the cylindrical section for the final homogenization step
- When pestle is “bottomed”- within the grinding chamber, the clearance is a uniform 0.102 to 0.152mm (0.004 to 0.006 in.)
Specifications
Borosilicate Glass | |
Grinding Chamber | |
Tapered tissue grinder with PTFE pestle | |
Tapered tissue grinder |