Learn More
DWK Life Sciences Wheaton™ 12 x 32 mm E-Z Vial Convenience Pack With 11 mm Crimp Finish And Clear Vial With Patch
Wheaton E-Z Vials have a 40% larger opening improves sample accessibility while reducing autosampler. Writing patch allows for identification needle damage.
Brand: DWK Life Sciences W225174-0203
Additional Details : Weight : 0.45360kg
Description
- Manufactured from certified clear Type I borosilicate glass
- Writing patch
- Vials are packaged 100 per shelf pack and 10 packs per case
- 40% larger opening improves sample accessibility while reducing autosampler needle damage
Specifications
Red | |
12 mm | |
1.8 mL | |
For chromatography | |
Aluminum | |
32 mm | |
11 mm |
Clear | |
Borosilicate Glass | |
PTFE, Silicone | |
1.25 in. | |
Crimp Cap | |
100 Cs. | |
Yes |