missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ White Polypropylene Caps for Narrow-Mouth Containers
Click to view available options
Neck Finish:
13-425
24-410
28-410
38-430
No. per Case:
48 Cs.
72 Cs.
Description
- Leak-proof design
- Autoclavable
Specifications
For Use With (Equipment) | Redi-pak™ leak resistant narrow mouth bottle |
Color | White |
Material | Polypropylene |
No. per Case | 72 Cs. |
Neck Finish | 13-425 |
Certifications/Compliance | ISO 9001 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'