Läs mer
3M™ White with Green Back Panel SMS Based Polypropylene Protective Coveralls
Handla allt 3M ProdukterSpecifikationer
Specifikationer
| Produkttyp | General Purpose Coveralls |
| Storlek | Small |
| Kön | Unisex |
| Disponibel | Single-use |
| Midja stil | Elastic |
| Manschettstil | Knit |
| Hals stil | Hooded |
| Huva typ | 3-panel |
| Sterilitet | Non-sterile |
| Skyddsklass | Category III |
| Visa mer |