Learn More
Fisherbrand™ White Polypropylene Caps
White polypropylene screw caps provides clear surface for writing on cap
Brand: Fisherbrand™
Description
- Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
- PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
- Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Specifications
No | |
Polypropylene | |
288 | |
White |
Pulp/Vinyl | |
White | |
Lined Cap | |
53-400 |