Läs mer
3M™ White with Red Seams Polypropylene/Polyethylene Laminate Film Protective Coveralls
Produktkod. 11957864 Handla allt 3M Produkter
Storlek:
Large
X-Large
Produktkod. 11957864
Brand: 3M™ 7000034774
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Specifikationer
Produkttyp General Purpose Coveralls
Storlek X-Large
Kön Unisex
Disponibel Single-use
Midja stil Elastic
Manschettstil Knit
Hals stil Hooded
Huva typ 3-panel
Sterilitet Non-sterile
Skyddsklass Category III
Höjd (metrisk) 179 to 187 cm