Läs mer
Bel-Art™ SP Scienceware™ Wilmad™ Aldrich FT-NMR Library with Spectral Viewer on CD-ROM
Handla allt SP Scienceware Produkter
Typ:
1,1800 Compound Database
3,500 Compound Database
Förpackningsstorlek:
Varje
Beskrivning
- Supplemental library that includes spectra for 3,500 additional compounds not included in the original printed version is also available
- Order both the standard and the supplemental library for access to spectra on over 15,000 compounds
- Multiple search methods: text Search by Chemical Name, Aldrich Catalog Number, CAS Number, Molecular Formula, advanced data field search on chemical categories and physical properties, such as Boiling Point, Melting Point, Density, Flash Point, Molecular Weight and Refractive Index
- Minimum System Requirement: Windows Platform, CD-ROM drive, 600MB hard disk space
Specifikationer
Specifikationer
| Typ | 3,500 Compound Database |
| Beskrivning | FT-NMR Library with Spectral Viewer on CD-ROM |