Läs mer
Bel-Art™ SP Scienceware™ WIlmad™ OmniFit™ Valve NMR System
Produktkod. 15328608
Change view
Modell:
507-AV-7
AV-30
OFV
Typ:
NMR-provrör
Skiljevägg
Ventil
Förpackningsstorlek:
Paket med 6
Varje
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Modell Typ unitSize
15328608 OFV Ventil Varje
15318608 AV-30 Skiljevägg Paket med 6
15398858 507-AV-7 NMR-provrör Varje
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 15328608 Leverantör Bel-Art™ Leverantörsnummer 660000038
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Designed for easy injection of chemicals through a gas-tight syringe for air sensitive samples without using a glove box. Bel-Art SP Scienceware™ WIlmad™ OmniFit™ Valve NMR System is comprised of a 5mm end cap, 2-way valve, septum with cap, barrel coupler and a vacuum connector.
  • The Omni-Fit Tube consists of a 507-PP tube topped by a sturdy 2 in. section of medium-walled tubing which supports the valve system
  • Omni-Fit Sample tubes (507-OF-7) sold separately
  • Omni-Fit valve (OFV) is a complete accessory set
  • Additional septum (OF-30) are available in packages of 6
Due to the nature of glass, Extreme Caution should be exercised when using at elevated or reduced pressures as performance of tubes under pressure or vacuum cannot be guaranteed. Adequate safety shielding should always be used when working in these conditions.
Specifikationer
Typ Ventil
Diameter (metrisk) 5.0 mm
Modell OFV