Läs mer
Bel-Art™ SP Scienceware™ Wilmad™ Universal Solvent Jet NMR Tube Washer
Handla allt SP Scienceware ProdukterBeskrivning
- Loosen and retighten the fitting on the assembled washer head to extend the PTFE tubing into the sample tube.
- Washer head affixes to a filter flask that includes a vacuum sidearm (sold separately)
- When a vacuum is applied to the flask, the sample tube is pressed against the rubber gasket to form an air-tight seal that starts the solvent flow.
- Sample tube can be air-dried by lifting the PTFE tubing out of the solvent reservoir
Specifikationer
Specifikationer
| Produkttyp | Single Tube Washer |
| Diameter (metrisk) Yttre | Fits tubing: 2.5 to 5.0mm O.D. |
| Inkluderar | Washer Body, rubber gasket, PTFE tubing, and PTFE tubing adapter. |
| Modell | WG-7200-1 |