missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIPI2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15432-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
WD repeat proteins play a role in many essential biologic functions, regulating the assembly of multiprotein complexes by presenting a beta-propeller platform for simultaneous and reversible protein-protein interactions. WIPI2, also known as ATG18B or ATG21, is a human homolog to yeast ATG18 and contains three WD repeats and has a 7-bladed propeller structure with a conserved motif that facilitates its interaction with other proteins. It is recruited to early autophagosomal structures along with Atg16L and ULK1 and is required for the formation of LC3-positive autophagosomes. Along with the highly related WIPI1, WIPI2 is found at the plasma membrane in addition to autophagosomal membranes.Specifications
WIPI2 | |
Polyclonal | |
Unconjugated | |
WIPI2 | |
Atg21, CGI 50, WIPI 2, WIPI2, WIPI49 like protein 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26100, 74781 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q80W47, Q9Y4P8 | |
Wipi2 | |
WIPI2 Fusion Protein Ag7689 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |