Läs mer
R&L Enterprises™ Wire Gauze with Ceramic Center
Produktkod. 12978950
Change view
Mått (L x B):
125 x 125 mm
Förpackningsstorlek:
Paket med 10
Produktkod. Mått (L x B) Förpackningsstorlek
12978950 125 x 125 mm Paket med 10
1 options
Produktkod. 12978950 Leverantör R&L Enterprises™ Leverantörsnummer 9072
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Provides excellent support to beakers and other glassware during heating on ring stands. R&L Enterprises Wire Gauze with Ceramic Center is a flat square manufactured from galvanized iron mesh.
  • Non-fraying edges
  • Durable and long-lasting
  • Ceramic center disperses heat evenly
Specifikationer
Produkttyp Wire Gauze
Mått (L x B) 125 x 125 mm
Inkluderar Ceramic center
Kvantitet 10 Pack
Längd (metrisk) 125 mm
Material Iron
Bredd (metrisk) 125 mm