Läs mer
Tocris Bioscience™ Withaferin A
:
1mg
Beskrivning
Withaferin A is a steroid lactone that displays anti-inflammatory, antitumor and antiangiogenic activity. Withaferin A inhibits endothelial cells (HUVEC) sprouting in vitro (IC50 = 12 nM) and in vivo, prevents NF-κB activation by inhibiting activation of IKKβ and inhibits chymotrypsin-like activity of the 20S proteasome. In an animal model of Huntington's disease (HD), Withaferin A delays disease progression and increases lifespan; in a cellular model it suppresses aggregation of mutant huntingtin, activates HSF1 and induces Hsp70 chaperone expression.
Specifikationer
Specifikationer
| Kvantitet | 1 mg |
| Produkttyp | Withaferin A |
| Renhet | >98% |
| Molekylvikt (g/mol) | 470.6 |
| Inhibitorer | Withaferin A |
For Research Use Only