Läs mer
Withanolide A, TRC
Produktkod. 30455312
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30455312 1 mg
1 options
Produktkod. 30455312 Leverantör Toronto Research Chemicals Leverantörsnummer TRCW4989501MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Withanolide A
CAS 32911-62-9
Molekylformel C28 H38 O6
Kvantitet 1 mg
Synonym (5α,6α,7α,22R)-6,7-epoxy-5,20,22-trihydroxy-1-oxo-ergosta-2,24-dien-26-oic Acid Δ-Lactone, (22R)-6α,7α-epoxy-5,20,22-trihydroxy-1-oxo-5α-Ergosta-2,24-dien-26-oic Acid Δ-Lactone, 6,7-Epoxy-1H-cyclopenta[a]phenanthrene Ergosta-2,24-dien-26-oic Acid Deriv., 5,20α-Dihydroxy-6α,7α-epoxy-1-oxowitha-2,24-dienolide, 5α,20αF(R)-Dihydroxy-6α,7α-epoxy-1-oxowitha-2,24-dienolide, Withaniol
InChI formel InChI=1S/C28H38O6/c1-14-13-20(33-24(30)15(14)2)27(5,31)18-9-8-16-21-17(10-12-25(16,18)3)26(4)19(29)7-6-11-28(26,32)23-22(21)34-23/h6-7,16-18,20-23,31-32H,8-13H2,1-5H3/t16-,17-,18-,20+,21-,22-,23-,25-,26-,27+,28-/m0/s1
LEDER CC1=C(C)C(=O)O[C@H](C1)[C@](C)(O)[C@H]2CC[C@H]3[C@@H]4[C@@H]5O[C@@H]5[C@@]6(O)CC=CC(=O)[C@]6(C)[C@H]4CC[C@]23C
Molekylvikt (g/mol) 470.6
Formel vikt 470.2668