Learn More
Socorex™ Work Station For Bulk Container
Brand:  Socorex™ 320.BC050
Product Code. 10734633
  • 3140.00 SEK / Each
Only null left
Estimated Shipment: 28-08-2025
Specifications
Workstation
50 mL
Work station for bulk container
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Socorex™ Work Station For Bulk Container > Capacity: 50mL