Learn More
Memmert™ Works Calibration Certificate for 3 Temperatures
Factory calibration certificate for 3 temperatures (+5/+37+60°C)
Brand: Memmert™ IPP110ecoD00129
Additional Details : Weight : 0.01100kg
Description
- +5, +37, +60 °C
- Accessory
- Factory calibration certificate for 3 temperatures
Specifications
IPP110eco | |
Works Calibration Certificate for 3 Temperatures |