Läs mer
Tocris Bioscience™ WZ 4002
Beskrivning
WZ 4002 is a potent irreversible mutant selective EGFR tyrosine kinase inhibitor. It blocks ATP-dependent autophosphorylation of EGFR carrying the T790M or L858R mutation (IC50 = 8 nM). WZ 4002 exhibits selectivity for mutant EGFR over wild-type. In mice bearing T790M mutant lung cancer cell lines WZ 4002 inhibits EGFR, AKT and ERK1/2 phosphorylation and results in significant tumor regression.
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-[3-[[5-Chloro-2-[[2-methoxy-4-(4-methyl-1-piperazinyl)phenyl]amino]-4-pyrimidinyl]oxy]phenyl]-2-propenamide |
| CAS | 1213269-23-8 |
| Kvantitet | 50 mg |
| Mål | EGF Receptor Inhibitors |
| Molekylformel | C25H27ClN6O3 |
| Renhet | 0.98 |