Läs mer
ABEBA™ X-Light White Occupational Shoes O2 ESD
Storlek:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Förpackningsstorlek:
1 par
Beskrivning
- Lightweight, comfortable white occupational shoes
- Comply to EN ISO 20347: 2012, O2 SRC
- ESD dissipative according EN 61340 class climatic 1
- Slip resistant Mono PU outsole SRC
- White technical fabric
- Silver ions lining
- Replaceable active comfort insole
Specifikationer
Specifikationer
| Färg | White |
| Material | Synthetic fabric |
| Storlek | 35 |
| Certifieringar/efterlevnad (Europa) | Comply to EN 20347: 2012 O2 SRC, ESD dissipative according EN 61340 |
| Kön | Unisex |
| Ankel stil | Low-cut |
| Vattentät | Water Resistant |
| Produkttyp | O2 SRC ESD |