Learn More
Draeger Safety X-Plore™ 2100 Particulate Half Mask
Draeger Safety X-Plore™ 2100 Particulate Half Mask is the reusable alternative to disposable dust masks.
Brand:  Draeger Safety R55915
Additional Details : Weight : 0.36000kg
Product Code. 15715563
  • 775.00 SEK / Each
Currently Unavailable: We have no information when this product will be available
View alternative products
Description
Description
  • Ideal solution for people working frequently in dusty environments
  • Replaceable filters provide protection against solid and liquid particles
  • Offers the perfect combination of comfort, safety and cost-effectiveness
Specifications
Specifications
Silicone
Medium/Large
5 P100 filters