Läs mer
Xamoterol Hemifumarate, TRC
Produktkod. 30455296
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30455296 10 mg
30455215 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30455296 Leverantör Toronto Research Chemicals Leverantörsnummer TRCX49980810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 73210-73-8
Molekylformel 2(C16H25N3O5) . C4H4O4
Molekylvikt (g/mol) 339.39
Synonym 1-(4-Hydroxyphenoxy)-3-[2-(4-morpholinocarboxamido)ethylamino]-2-propanol Hemifumarate
IUPAC-namn (E)-but-2-enedioic acid;N-[2-[[2-hydroxy-3-(4-hydroxyphenoxy)propyl]amino]ethyl]morpholine-4-carboxamide
LEDER OC(CNCCNC(=O)N1CCOCC1)COc2ccc(O)cc2.OC(CNCCNC(=O)N3CCOCC3)COc4ccc(O)cc4.OC(=O)\C=C\C(=O)O
Specifikationer
Kemiskt namn eller material Xamoterol Hemifumarate
Kvantitet 10 mg
InChI formel InChI=1S/2C16H25N3O5.C4H4O4/c2*20-13-1-3-15(4-2-13)24-12-14(21)11-17-5-6-18-16(22)19-7-9-23-10-8-19;5-3(6)1-2-4(7)8/h2*1-4,14,17,20-21H,5-12H2,(H,18,22);1-2H,(H,5,6)(H,7,8)/b;;2-1+
Formel vikt 794.3698