Läs mer
Xanthopterin Hydrate, TRC
Produktkod. 30456298
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30456298 250 mg
30452175 100 mg
30452515 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30456298 Leverantör Toronto Research Chemicals Leverantörsnummer TRCX742003250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5979-01-1
Molekylformel C6H5N5O2 . H2O
Synonym 2-Amino-4,6-pteridinediol Hydrate
IUPAC-namn 2-amino-1,5-dihydropteridine-4,6-dione;hydrate
LEDER O.NC1=NC(=O)C2=C(N1)N=CC(=O)N2
Specifikationer
Kemiskt namn eller material Xanthopterin Hydrate
Kvantitet 250 mg
InChI formel InChI=1S/C6H5N5O2.H2O/c7-6-10-4-3(5(13)11-6)9-2(12)1-8-4;/h1H,(H,9,12)(H3,7,8,10,11,13);1H2
Formel vikt 197.0549