Learn More
Trajan Scientific and Medical™ XCHANGE™ CTC PAL-XT Syringe for GC
The knowledge and expertise that goes into manufacturing an SGE syringe gives outstanding performance and superior quality.
Brand:  Trajan Scientific and Medical™ 2928710
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15249273
  • 1140.00 SEK / Each
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
Specifications
Specifications
CTC PAL-XT Syringe
0.15mm
500μL
56mm
23g
0.63mm
For gas chromatography, liquid handling
Cone
Videos