Läs mer
XEN445, MedChemExpress
Produktkod. 16481277
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16481277 50 mg 50mg
16467767 5 mg -
16445786 100 mg -
16415048 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16481277 Leverantör Medchem Express Leverantörsnummer HY12246
MedChemExpress XEN445 is a potent and selective EL inhibitor(IC50=0.237 uM), that showed good ADME and PK properties, and demonstrated in vivo efficacy in raising plasma HDLc concentrations in mice.
XEN445 is a potent and selective EL inhibitor(IC50=0.237 uM), that showed good ADME and PK properties, and demonstrated in vivo efficacy in raising plasma HDLc concentrations in mice.
Specifikationer
Kemiskt namn eller material XEN445
Fysisk form Solid
CAS 1515856-92-4
Färg Light Brown to Black
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C18H17F3N2O3
För användning med (applikation) COVID-19-immunoregulation
Löslighetsinformation DMSO : ≥ 40 mg/mL (109.19 mM)
LEDER O=C(O)C1=CC(C(F)(F)F)=CC=C1N2C[C@@H](OCC3=NC=CC=C3)CC2
Molekylvikt (g/mol) 366.33
Formel vikt 366.33
Procent renhet 95.09%