Learn More
Uvex Xenova™ Black/Green PU 95643 Low Shoe - Width 12
Uvex 2 Xenova™ design provides comfortable, metal-free safety shoes; light weight with optimum cushioning and breathability for indoor and outdoor work
1320.00 SEK
Specifications
Color | Black, Green |
---|---|
Material | Leather |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17288741
|
Uvex
9564338 |
38 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17298741
|
Uvex
9564339 |
39 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17208751
|
Uvex
9564340 |
40 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17218751
|
Uvex
9564341 |
41 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17228751
|
Uvex
9564342 |
42 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17238751
|
Uvex
9564343 |
43 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17248751
|
Uvex
9564344 |
44 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17258751
|
Uvex
9564345 |
45 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17268751
|
Uvex
9564346 |
46 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17278751
|
Uvex
9564347 |
47 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17288751
|
Uvex
9564348 |
48 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17298751
|
Uvex
9564349 |
49 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17208761
|
Uvex
9564350 |
50 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17218761
|
Uvex
9564351 |
51 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17228761
|
Uvex
9564352 |
52 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Uvex 2 Xenova™ Shoe 95643 S3 SRC
- Extra wide sports fit
- Latest generation of 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good side stability, no thermal conductivity and anti-magnetic properties
- Penetration-resistant, non-metallic midsole in accordance with latest safety standards, does not restrict flexibility
- Breathable, water repellent full grain leather
- Scratch-resistant scuffcap made from PU coated leather for longer service life
- Distance mesh lining provides optimum breathability
- Made from synthetic textiles, suitable for those allergic to chrome
- Uvex anti-twist heel counter for more stability and prevention of twisting ankle
- Soft, padded collar and dust tongue
- Comfortable removable insole
- Antistatic with a moisture control system
- Double density PUR sole, non-marking, with excellent shock absorption and abrasion resistant outsole
- Self-cleaning tread design
- Oil and fuel resistant, free of lacquer wetting inhibitors such as silicones, softeners, etc.
- Short term heat resistant up to 120°C
- Antistatic
- Fulfills ESD requirements: resistance to ground Kleiner to 35 megohms
- Article No. 95643
- Standard EN ISO 20345:2011 S3 SRC
- Sizes: 38 to 52
Specifications
Black, Green | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Leather | |
Composite | |
EN ISO 20345:2011 | |
Wide width | |
Yes | |
S3 |