missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPNPEP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | XPNPEP2 |
---|---|
Concentration | 0.2 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16824485
|
Proteintech
25945-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16814485
|
Proteintech
25945-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Aminopeptidase P is a hydrolase specific for N-terminal imido bonds, which are common to several collagen degradation products, neuropeptides, vasoactive peptides, and cytokines. Structurally, the enzyme is a member of the ′pita bread fold′ family and occurs in mammalian tissues in both soluble and GPI-anchored membrane-bound forms. A membrane-bound and soluble form of this enzyme have been identified as products of two separate genes.Specifications
XPNPEP2 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
O43895 | |
7512 | |
XPNPEP2 Fusion Protein Ag22952 | |
Primary | |
-20°C | |
XPNPEP2 |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
mAmP, Membrane bound AmP, Membrane bound APP, X Pro aminopeptidase 2, Xaa Pro aminopeptidase 2, XPNPEP2 | |
XPNPEP2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |