Learn More
Abnova™ XPO1 Recombinant Protein
Human XPO1 full-length ORF (AAH32847.1, 1 a.a. - 1071 a.a.) recombinant protein with GST-tag at N-terminal
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH32847.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 7514 |
Molecular Weight (g/mol) | 143.55 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16151902
|
Abnova™
H00007514-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16161902
|
Abnova™
H00007514-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene mediates leucine-rich nuclear export signal (NES)-dependent protein transport. Exportin 1 specifically inhibits the nuclear export of Rev and U snRNAs. It is involved in the control of several cellular processes by controlling the localization of cyclin B, MPAK, and MAPKAP kinase 2. This protein also regulates NFAT and AP-1.
- Exportin 1 (CRM1 homolog, yeast)
- Molecular weight: 143.55kDa
- Preparation method: in vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 Fast Flow
- Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8 in the elution buffer
- Quality Control Testing: 12.5% SDS-PAGE stained with Coomassie Blue
Sequence:
MPAIMTMLADHAARQLLDFSQKLDINLLDNVVNCLYHGEGAQQRMAQEVLTHLKEHPDAWTRVDTILEFSQNMNTKYYGLQILENVIKTRWKILPRNQCEGIKKYVVGLIIKTSSDPTCVEKEKVYIGKLNMVLVQILKQEWPKHWPTFISDIVGASRTSESPCQNNMVILKLLSEEVFDFSSGQITQVKSKHLKDSMCNEFSQIFQLCQFVMENSQNAPLVHATLETLLRFLNWIPLGYIFETKLISTLIYKFLNVPMFRNVSLKCLTEIAGVSVSQYEEQFVTLFTLTMMQLKQMLPLNTNIRLAYSNGKDDEQNFIQNLSLFLCTFLKEHDQLIEKRLNLRETLMEALHYMLLVSEVEETEIFKICLEYWNHLAAELYRESPFSTSASPLLSGSQHFDVPPRRQLYLPMLFKVRLLMVSRMAKPEEVLVVENDQGEVVREFMKDTDSINLYKNMRETLVYLTHLDYVDTERIMTEKLHNQVNGTEWSWKNLNTLCWAIGSISGAMHEEDEKRFLVTVIKDLLGLCEQKRGKDNKAIIASNIMYIVGQYPRFLRAHWKFLKTVVNKLFEFMHETHDGVQDMACDTFIKIAQKCRRHFVQVQVGEVMPFIDEILNNINTIICDLQPQQVHTFYEAVGYMIGAQTDQTVQEHLIEKYMLLPNQVWDSIIQQATKNVDILKDPETVKQLGSILKTNVRACKAVGHPFVIQLGRIYLDMLNVYKCLSENISAAIQANGEMVTKQPLIRSMRTVKRETLKLISGWVSRSNDPQMVAENFVPPLLDAVLIDYQRNVPAAREPEVLSTMAIIVNKLGGHITAEIPQIFDAVFECTLNMINKDFEEYPEHRTNFFLLLQAVNSHCFPAFLAIPPTQFKLVLDSIIWAFKHTMRNVADTGLQILFTLLQNVAQEEAAAQSFYQTYFCDILQHIFSVVTDTSHTAGLTMHASILAYMFNLVEEGKISTSLNPGNPVNNQIFLQEYVANLLKSAFPHLQDAQVKLFVTGLFSLNQDIPAFKEHLRDFLVQIKEFAGEDTSDLFLEEREIALRQADEEKHKRQMSVPGIFNPHEIPEEMCD
Best use within three months from the date of receipt of this protein.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH32847.1 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
143.55 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
XPO1 | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
7514 | |
XPO1 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPAIMTMLADHAARQLLDFSQKLDINLLDNVVNCLYHGEGAQQRMAQEVLTHLKEHPDAWTRVDTILEFSQNMNTKYYGLQILENVIKTRWKILPRNQCEGIKKYVVGLIIKTSSDPTCVEKEKVYIGKLNMVLVQILKQEWPKHWPTFISDIVGASRTSESPCQNNMVILKLLSEEVFDFSSGQITQVKSKHLKDSMCNEFSQIFQLCQFVMENSQNAPLVHATLETLLRFLNWIPLGYIFETKLISTLIYKFLNVPMFRNVSLKCLTEIAGVSVSQYEEQFVTLFTLTMMQLKQMLPLNTNIRLAYSNGKDDEQNFIQNLSLFLCTFLKEHDQLIEKRLNLRETLMEALHYMLLVSEVEETEIFKICLEYWNHLAAELYRESPFSTSASPLLSGSQHFDVPPRRQLYLPMLFKVRLLMVSRMAKPEEVLVVENDQGEVVREFMKDTDSINLYKNMRETLVYLTHLDYVDTERIMTEKLHNQVNGTEWSWKNLNTLCWAIGSISGAMHEEDEKRFLVTVIKDLLGLCEQKRGKDNKAIIASNIMYIVGQYPRFLRAHWKFLKTVVNKLFEFMHETHDGVQDMACDTFIKIAQKCRRHFVQVQVGEVMPFIDEILNNINTIICDLQPQQVHTFYEAVGYMIGAQTDQTVQEHLIEKYMLLPNQVWDSIIQQATKNVDILKDPETVKQLGSILKTNVRACKAVGHPFVIQLGRIYLDMLNVYKCLSENISAAIQANGEMVTKQPLIRSMRTVKRETLKLISGWVSRSNDPQMVAENFVPPLLDAVLIDYQRNVPAAREPEVLSTMAIIVNKLGGHITAEIPQIFDAVFECTLNMINKDFEEYPEHRTNFFLLLQAVNSHCFPAFLAIPPTQFKLVLDSIIWAFKHTMRNVADTGLQILFTLLQNVAQEEAAAQSFYQTYFCDILQHIFSVVTDTSHTAGLTMHASILAYMFNLVEEGKISTSLNPGNPVNNQIFLQEYVANLLKSAFPHLQDAQVKLFVTG | |
CRM1/DKFZp686B1823 | |
XPO1 | |
Wheat Germ (in vitro) | |
GST |