Läs mer
Tocris Bioscience™ XY1
GREENER_CHOICE
Negative control of SGC 707 (Cat. No. 5367)
Brand:  Tocris Bioscience™ 5450/10
Encompass
Produktkod. 15736846
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
XY1 is a negative control of SGC 707 . Exhibits >3000-fold lower potency (IC50 >100 μM) compared to the active analog.
Active Analog also available.
Specifikationer
Specifikationer
>98%
10 mg
Produktförslag
Produktförslag
Filmer
SDS
Dokument
Dokument
Certifikat
Kampanjer
Kampanjer