Läs mer
Xylan, MedChemExpress
Beskrivning
Xylan represents the main hemicellulose component in the secondary plant cell walls of flowering plants. Xylan is a polysaccharide made from units of xylose and contains predominantly β-D-xylose units linked as in cellulose.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Xylan |
| Fysisk form | Solid |
| CAS | 9014-63-5 |
| Färg | White |
| Kvantitet | 1 g |
| Anteckningar om renhetsgrad | Research |
| Löslighetsinformation | H2O : 110 mg/mL (Need ultrasonic) ∣DMSO : 100 mg/mL (Need ultrasonic) |
| LEDER | [Xylan] |
| Kvalitet | Research |