missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XYLB Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26541-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Human xylulokinase-like protein (XYLB), is a 58 kDa protein belongs to a family of enzymes that include fucokinase, gluconokinase, glycerokinase and xylulokinase. XYLB is a key regulator for controlling metabolic disease. XYLB also plays important roles in energy metabolism. XYLB has 2isoformsproduced byalternative splicing with the MW of 58 and 43 kDa.Specifications
XYLB | |
Polyclonal | |
Unconjugated | |
XYLB | |
XYLB, Xylulokinase, Xylulose kinase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9942 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75191 | |
XYLB | |
XYLB Fusion Protein Ag19208 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |