Läs mer
Xylene, Solveco
Beskrivning
Premium-grade xylene with high and consistent purity, specially developed for pathological and cytological applications. Its excellent solvency power ensures fast and thorough deparaffinization and effective cleaning of tissue samples, contributing to reliable and reproducible staining results.
Ideal for use in histology and cytology labs, this xylene is also suitable for various industrial applications as a solvent and thinner, similar to toluene. It is commonly used in coatings, adhesives, and chemical manufacturing processes where precision and purity are essential.
- High purity and batch consistency
- Optimized for tissue processing and staining
- Rapid and complete deparaffinization
- Also suitable as industrial solvent and thinner
Packaged to meet the needs of both laboratory and industrial environments.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Dimetylbensen |
| Kvantitet | 25 L |
| Förpackning | 25 L HDPE Container |
| Kokpunkt | 137 to 143°C |
| Kvalitet | Technical |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.