Learn More
Xylenes, ≥98.5%, ACS Reagent, xylenes plus ethylbenzene basis, Honeywell™ Riedel-de Haën™
Catalog Number p-7120902
Click to view available options
Quantity:
500 mL
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
1330-20-7
MFCD00077264
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
1330-20-7
MFCD00077264
ACS Reagent,≥98.5% xylenes + ethylbenzene basis
ACS GENERAL USE SOLVENTS
  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY
Melting Point -48°C
Linear Formula C6H4(CH3)2
UN Number UN1307
Formula Weight 106.17g/mol
Percent Purity ≥98.5% xylenes plus ethylbenzene basis
Grade ACS Reagent
Density 0.86g/mL at 25°C
Boiling Point 137°C to 140°C
Quantity 500 mL
Explosion Limit 0.07
Flash Point 25°C (77°F)
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 18mmHg (37.7°C)
Chemical Name or Material Xylenes