missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, contains ethylbenzene, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5383264
Click to view available options
Quantity:
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
1330-20-7
C6H4(CH3)2
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
1330-20-7
C6H4(CH3)2

High quality chemicals for laboratory and specialized industrial use

    For Microelectronic Use
Formula Weight 106.17
Grade BAKER ANALYZED™ ACS Reagent
Quantity 4 L
Flash Point 29.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.86kg
Chemical Name or Material Xylenes, Contains ethylbenzene
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'