Läs mer
Xylenesulfonic Acid Sodium Salt, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Xylenesulfonic Acid Sodium Salt |
| CAS | 1300-72-7 |
| Molekylformel | C8H9NaO3S |
| Kvantitet | 25 g |
| Synonym | Dimethylbenzenesulfonic Acid Sodium Salt, Conco SXS, Cyclophil SXS 30, Eltesol SX 30, Eltesol SX 93, FAC 2, Kemmat SN 18, Naxonate, Naxonate 4L, Naxonate G, Naxonate SX, Richonate SXS, SXS 40, Sodium Dimethylbenzenesulfonate, Sodium Xylenesulfate, Sodium Xylenesulfonate, Stepanate SXS, Stepanate X, Ultrawet 40SX, Vanwet SXS 40, Witconate SXS |
| InChI formel | InChI=1S/C8H10O3S.Na/c1-6-3-4-8(5-7(6)2)12(9,10)11;/h3-5H,1-2H3,(H,9,10,11);/q;+1/p-1 |
| LEDER | [Na+].Cc1ccc(cc1C)S(=O)(=O)[O-] |
| Molekylvikt (g/mol) | 208.21 |