Läs mer
Xylobiose, TRC
Produktkod. 30456230
Change view
Kvantitet:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30456230 10 mg
30451646 5 mg
30452177 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30456230 Leverantör Toronto Research Chemicals Leverantörsnummer TRCX75040010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6860-47-5
Molekylformel C10H18O9
Molekylvikt (g/mol) 282.24
Synonym 1,4-beta-Xylobiose, 4-O-beta-D-Xylopyranosyl-D-xylose
IUPAC-namn (2R,3R,4R)-2,3,5-trihydroxy-4-[(2S,3R,4S,5R)-3,4,5-trihydroxyoxan-2-yl]oxypentanal
LEDER OC[C@@H](O[C@@H]1OC[C@@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O
Specifikationer
Kemiskt namn eller material Xylobiose
Kvantitet 10 mg
InChI formel InChI=1S/C10H18O9/c11-1-4(13)7(15)6(2-12)19-10-9(17)8(16)5(14)3-18-10/h1,4-10,12-17H,2-3H2/t4-,5+,6+,7+,8-,9+,10-/m0/s1
Formel vikt 282.0951