Läs mer
Xylotriose, TRC
Produktkod. 30456463
Change view
Kvantitet:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30456463 100 mg
30453786 5 mg
30451928 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30456463 Leverantör Toronto Research Chemicals Leverantörsnummer TRCX750405100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 47592-59-6
Molekylformel C15H26O13
Molekylvikt (g/mol) 414.36
Synonym O-β-D-Xylopyranosyl-(1→4)-O-β-D-xylopyranosyl-(1→4)-D-xylose
IUPAC-namn (2S,3R,4S,5R)-2-[(3R,4R,5R,6S)-4,5-dihydroxy-6-[(3R,4R,5R)-4,5,6-trihydroxyoxan-3-yl]oxyoxan-3-yl]oxyoxane-3,4,5-triol
LEDER O[C@H]1[C@H](O)[C@@H](O)[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@H]3[C@H](O)[C@@H](O)C(O)OC3)OC2)OC1
Specifikationer
Kemiskt namn eller material Xylotriose
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C15H26O13/c16-4-1-25-14(11(21)7(4)17)28-6-3-26-15(12(22)9(6)19)27-5-2-24-13(23)10(20)8(5)18/h4-23H,1-3H2/t4-,5-,6-,7+,8+,9+,10-,11-,12-,13?,14+,15+/m1/s1
Formel vikt 414.1373
Procent renhet >95%