Läs mer
Tocris Bioscience™ YADA
Beskrivning
YADA is a green-yellow lucifer yellow-based fluorescent D-amino acid. Suitable for labeling peptidoglycans in live bacteria. Incorporated into bacterial cell walls during synthesis. Exhibits large stokes shift and wider emission spectra than other FDAAs, allowing excitation via a violet light source and detection with a green filter. Displays good water solubility, photostability and thermostability. Excitation/emission λ ∽426/535 nm.
Specifikationer
Specifikationer
| Kemiskt namn eller material | (αR)-α,6-Diamino-1,3-dioxo-5,8-disulfo-1 H-benz[de]isoquinoline-2(3 H)-propanoic acid |
| CAS | 1471982-33-8 |
| Kvantitet | 5 mg |
| Mål | Fluorescent Probes for Imaging Bacteria |
| Molekylformel | C15H13N3O10S2 |
| Formel vikt | 3636 |
| Renhet | 95% |