Läs mer
Tocris Bioscience™ YE 120
Produktkod. 18075402
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18075402 10 mg
18095632 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18075402 Leverantör Tocris Bioscience™ Leverantörsnummer 4457/10
GPR35 agonist
GPR35 agonist (EC50 = 32 nM in DMR assays). Also displays partial agonist activity in a β-arrestin translocation assay (EC50 = 10.2 μM).
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 383124-82-1
Molekylformel C16H9Cl2N3O
Specifications
Kemiskt namn eller material YE 120
Kvantitet 10 mg
Formel vikt 330.17
Renhet >98%
Molekylvikt (g/mol) 330.168