Learn More
Eppendorf™ Yellow Color Code Counter Shutters
Designed for use with Eppendorf Pipettes
Brand:  Eppendorf™ 3121804009
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 15274376
  • 43.00 SEK / Each
Estimated Shipment
04-09-2018
Add to basket
Description
Description
  • Accessory for Eppendorf pipettes
  • For use with liquid handling applications
  • Counter shutters available for fixed and variable pipettes
Specifications
Specifications
For 10μL Single-channel Fixed Pipette
1Each
Color Code Counter Shutter
Videos